2,5-Dichlorothiophene ≥98%

Supplier: Matrix Scientific

005624-100G 005624-25G
101778-550EA 52.31 USD
101778-550 101778-548
2,5-Dichlorothiophene ≥98%
2,5-Dichlorothiophene

TCA listed: T
Hazard: Flammable

Formula: C₄H₂Cl₂S
MW: 153.03 g/mol
Boiling Pt: 162 °C
Density: 1.442
MDL Number: MFCD00005423
CAS Number: 3172-52-9
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 162°
Density 1.442
ND20 1.562

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR