(3,4-Dichlorophenyl)acetic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
3,4-Dichlorophenylacetic acid
005576-100G
005576-25G
101778-502EA
21.54
USD
101778-502
101778-504
(3,4-Dichlorophenyl)acetic acid ≥98%
(3,4-Dichlorophenyl)acetic acid
Hazard: Irritant
Formula:
(C₆H₃Cl₂)CH₂COOH MW: 205.04 g/mol Melting Pt: 89…91 °C |
MDL Number:
MFCD00004333 CAS Number: 5807-30-7 |
Specification Test Results
Melting point | 89-91° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...