4-(Trifluoromethylthio)phenyl isocyanate
Supplier: Matrix Scientific
005540-1G
101778-466EA
144.62
USD
101778-466
4-(Trifluoromethylthio)phenyl isocyanate
4-(Trifluoromethylthio)phenyl isocyanate
Hazard: Toxic/Store in Freezer/Lachrymator
Formula:
C₈H₄F₃NOS MW: 219.19 g/mol Boiling Pt: 43 °C (0.01 mmHg) Density: 1.365 |
MDL Number:
MFCD00036025 CAS Number: 24032-84-6 |
Specification Test Results
Boiling point | 43°/0.01mm |
Density | 1.365 |
ND20 | 1.511 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...