3-(Trifluoromethylthio)phenylacetonitrile

Supplier: Matrix Scientific

005537-1G
101778-464EA 46 USD
101778-464
3-(Trifluoromethylthio)phenylacetonitrile
3-(Trifluoromethylthio)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₆F₃NS
MDL Number: MFCD00236353
CAS Number: 82174-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR