2-(Trifluoromethylthio)phenylacetonitrile

Supplier: Matrix Scientific

005536-250MG
101778-462EA 79.35 USD
101778-462
2-(Trifluoromethylthio)phenylacetonitrile
2-(Trifluoromethylthio)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₆F₃NS
MDL Number: MFCD00236352
CAS Number: 237424-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR