2-(Trifluoromethyl)acrylic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)propenoic acid
005505-25G
005505-5G
101778-424EA
64.62
USD
101778-424
101778-426
2-(Trifluoromethyl)acrylic acid ≥98%
2-(Trifluoromethyl)acrylic acid
Hazard: Corrosive
Formula:
C₄H₃F₃O₂ MW: 140.06 g/mol Boiling Pt: 86 °C (35 mmHg) |
MDL Number:
MFCD00042424 CAS Number: 381-98-6 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 86°/35mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...