4-(Trifluoromethyl)phenyl isothiocyanate ≥99%

Supplier: Matrix Scientific

Synonyms: α,α,α-Trifluoro-p-tolyl isothiocyanate1-Isocyanato-4-(trifluoromethyl)benzene

005503-5G
101778-422EA 81.54 USD
101778-422
4-(Trifluoromethyl)phenyl isothiocyanate ≥99%
4-(Trifluoromethyl)phenyl isothiocyanate

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 81 °C (11 mmHg)
Melting Pt: 41…43 °C
MDL Number: MFCD00039645
CAS Number: 1645-65-4
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Boiling point 81°/11mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR