α'-Chloro-α,α,α-trifluoro-p-xylene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)benzyl chloride4-(Chloromethyl)benzotrifluoride1-(Chlormethyl)-4-(trifluormethyl)benzene1-(Chloromethyl)-4-(trifluoromethyl)benzene

005491-100G 005491-25G
101778-400EA 53.85 USD
101778-400 101778-402
α'-Chloro-α,α,α-trifluoro-p-xylene ≥98%
α'-Chloro-α,α,α-trifluoro-p-xylene

Hazard: Corrosive/Lachrymator

Formula: C₈H₆ClF₃
MW: 194.58 g/mol
Boiling Pt: 68 °C (12 mmHg)
Density: 1.315
MDL Number: MFCD00040772
CAS Number: 939-99-1
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Boiling point 68°/12mm
Density 1.315
ND20 1.464

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR