2-(Trifluoromethoxy)benzoyl chloride ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethoxy)benzene-1-carbonyl chloride
005377-25G
005377-5G
101778-320EA
29.23
USD
101778-320
101778-322
2-(Trifluoromethoxy)benzoyl chloride ≥98%
2-(Trifluoromethoxy)benzoyl chloride
Hazard: Corrosive/Refrigerate
Formula:
C₈H₄ClF₃O₂ MW: 224.57 g/mol Boiling Pt: 94…96 °C (15 mmHg) |
MDL Number:
MFCD00052330 CAS Number: 162046-61-9 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 94-96°/15m |
ND20 | 1.47 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...