4-(Trifluoromethoxy)phenol ≥98%

Supplier: Matrix Scientific

Synonyms: α,α,α-Trifluoro-4-hydroxyanisole

005374-100G
101778-318EA 75.38 USD
101778-318
4-(Trifluoromethoxy)phenol ≥98%
4-(Trifluoromethoxy)phenol

Hazard: Toxic/Irritant

Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
Boiling Pt: 92 °C (25 mmHg)
Density: 1.375
MDL Number: MFCD00040988
CAS Number: 828-27-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 92°/25mm
Density 1.375
ND20 1.447

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR