3-(Trifluoromethoxy)anisole

Supplier: Matrix Scientific

Synonyms: 1-Methoxy-3-(trifluoromethoxy)benzene

005340-1G 005340-5G
101778-280EA 13.85 USD
101778-280 101778-282
3-(Trifluoromethoxy)anisole
3-(Trifluoromethoxy)anisole

Hazard: Irritant

Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
MDL Number: MFCD01631504
CAS Number: 142738-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR