α-Bromo-4-(trifluoromethoxy)toluene ≥97%
Supplier: Matrix Scientific
Synonyms:
p-(Trifluoromethoxy)benzyl bromide
, 4-(Trifluoromethoxy)benzyl bromide
005336-100G
101778-278EA
287.69
USD
101778-278
α-Bromo-4-(trifluoromethoxy)toluene ≥97%
α-Bromo-4-(trifluoromethoxy)toluene
Hazard: Corrosive/Refrigerate/Lachrymator
Formula:
C₈H₆BrF₃O MW: 255.03 g/mol Boiling Pt: 63 °C (3 mmHg) Melting Pt: 22…24 °C Density: 1.594 |
MDL Number:
MFCD00061238 CAS Number: 50824-05-0 UN: 3261 ADR: 8,III |
Specification Test Results
Boiling point | 63°/3mm |
Density | 1.594 |
ND20 | 1.48 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...