2-(Trifluoromethoxy)phenyl isothiocyanate

Supplier: Matrix Scientific

005334-1G
101778-274EA 36.92 USD
101778-274
2-(Trifluoromethoxy)phenyl isothiocyanate
2-(Trifluoromethoxy)phenyl isothiocyanate

Hazard: Irritant

Formula: C₈H₄F₃NOS
MW: 219.19 g/mol
MDL Number: MFCD00220860
CAS Number: 175205-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR