3-(Trifluoromethoxy)benzoyl chloride ≥97%

Supplier: Matrix Scientific

005318-25G 005318-5G
101778-230EA 90.77 USD
101778-230 101778-232
3-(Trifluoromethoxy)benzoyl chloride ≥97%
3-(Trifluoromethoxy)benzoyl chloride

Hazard: Corrosive/Refrigerate

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
MDL Number: MFCD00061268
CAS Number: 86270-03-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR