1,1-Difluoroethylene ≥97%
Supplier: Matrix Scientific
005276-1KG
005276-100G
101778-200EA
684.62
USD
101778-200
101778-202
1,1-Difluoroethylene ≥97%
1,1-Difluoroethylene
TCA listed: T
Hazard: Flammable Gas
Formula:
C₂H₂F₂ MW: 64.03 g/mol Boiling Pt: –83 °C Melting Pt: –144 °C |
MDL Number:
MFCD00000448 CAS Number: 75-38-7 UN: 1959 ADR: 2, |
Specification Test Results
Boiling point | -83° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...