Undecafluorocyclohexanecarboxylic acid ≥97%

Supplier: Matrix Scientific

005259-1G 005259-5G
101778-176EA 36.92 USD
101778-176 101778-178
Undecafluorocyclohexanecarboxylic acid ≥97%
Undecafluorocyclohexanecarboxylic acid

Hazard: Corrosive

Formula: C₇HF₁₁O₂
Boiling Pt: 168…170 °C (742 mmHg)
Density: 1.78
MDL Number: MFCD01075297
CAS Number: 374-88-9

Order Now

Specification Test Results

Boiling point 168-170°/7

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR