α-Bromo-3,4-difluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)-1,2-difluorobenzene3,4-Difluorobenzyl bromide

005235-25G 005235-100G
101778-126EA 60 USD
101778-126 101778-128
α-Bromo-3,4-difluorotoluene ≥97%
α-Bromo-3,4-difluorotoluene

Hazard: Corrosive/Irritant/Lachrymator

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Density: 1.618
MDL Number: MFCD00009889
CAS Number: 85118-01-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.618
ND20 1.526

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR