1,4-Difluorobenzene ≥99%

Supplier: Matrix Scientific

005187-500G 005187-100G
101778-076EA 283.08 USD
101778-076 101778-074
1,4-Difluorobenzene ≥99%
1,4-Difluorobenzene

Hazard: Flammable

Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 88…89 °C
Melting Pt: –13 °C
Density: 1.11
MDL Number: MFCD00000344
CAS Number: 540-36-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 88-89°
Density 1.11
ND20 1.441

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR