2,6-Difluorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluorobenzene-1-sulfonyl chloride2,6-Difluorobenzene-1-sulphonyl chloride

005183-25G
101778-062EA 60 USD
101778-062
2,6-Difluorobenzenesulfonyl chloride ≥98%
2,6-Difluorobenzenesulphonyl chloride

Hazard: Corrosive

Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 105…110 °C (2 mmHg)
MDL Number: MFCD00085004
CAS Number: 60230-36-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 105-110°/2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR