2,5-Difluoroaniline ≥99%

Supplier: Matrix Scientific

005147-100G 005147-25G
101777-990EA 72.31 USD
101777-990 101777-988
2,5-Difluoroaniline ≥99%
2,5-Difluoroaniline

Hazard: Toxic/Irritant

Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 176…178 °C
Melting Pt: 13…14 °C
Density: 1.288
MDL Number: MFCD00007651
CAS Number: 367-30-6
UN: 2941
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 176-178°
Density 1.288
ND20 1.513

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR