3,5-Difluoroaniline ≥98%

Supplier: Matrix Scientific

005144-100G
101777-976EA 129.23 USD
101777-976
3,5-Difluoroaniline ≥98%
3,5-Difluoroaniline

Hazard: Toxic/Irritant

Formula: C₆H₅F₂N
MW: 129.11 g/mol
Melting Pt: 40…42 °C
MDL Number: MFCD00007763
CAS Number: 372-39-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 40-42°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR