α-Bromo-2,4-dichloro-5-fluorotoluene
Supplier: Matrix Scientific
005107-5G
005107-1G
101777-942EA
264.62
USD
101777-942
101777-940
α-Bromo-2,4-dichloro-5-fluorotoluene
α-Bromo-2,4-dichloro-5-fluorotoluene
Hazard: Corrosive
Formula:
C₇H₄BrCl₂F MW: 257.92 g/mol Boiling Pt: 94 °C (1 Torr) |
CAS Number:
261763-27-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...