(2,3-Difluoro-4-(trifluoromethyl)phenyl)acetonitrile
Supplier: Matrix Scientific
005040-250MG
005040-1G
101777-918EA
315.38
USD
101777-918
101777-916
(2,3-Difluoro-4-(trifluoromethyl)phenyl)acetonitrile
(2,3-Difluoro-4-(trifluoromethyl)phenyl)acetonitrile
Hazard: Toxic
Formula:
C₉H₄F₅N |
MDL Number:
MFCD00236270 CAS Number: 240800-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...