2,6-Difluoro-3-methylbenzyl bromide

Supplier: Matrix Scientific

005019-1G
101777-906EA 49.23 USD
101777-906
2,6-Difluoro-3-methylbenzyl bromide
2,6-Difluoro-3-methylbenzyl bromide

Hazard: Corrosive

Formula: C₈H₇BrF₂
MW: 221.04 g/mol
Boiling Pt: 76 °C (3 Torr)
CAS Number: 261763-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR