2,6-Difluoro-3-methylbenzylamine

Supplier: Matrix Scientific

005018-1G
101777-904EA 83.95 USD
101777-904
2,6-Difluoro-3-methylbenzylamine
2,6-Difluoro-3-methylbenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₉F₂N
MDL Number: MFCD01631410
CAS Number: 261763-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR