2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride

Supplier: Matrix Scientific

004974-5G 004974-1G
101777-876EA 392.31 USD
101777-876 101777-874
2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride
2,2-Difluoro-1,3-benzodioxole-4-carbonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₃ClF₂O₃
MDL Number: MFCD01631474
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR