Hexafluoroglutaryl fluoride ≥97%

004881-1G 004881-5G
101777-814EA 1653.5 USD
101777-814 101777-816
Hexafluoroglutaryl fluoride ≥97%
Hexafluoroglutaryl fluoride

Hazard: Corrosive/Store in Freezer

Formula: C₅F₈O₂
Boiling Pt: 46…47 °C
Density: 1.6389
MDL Number: MFCD00054652
CAS Number: 678-78-4

Order Now

Specification Test Results

Boiling point 46-47°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR