Hexafluoroacetone sesquihydrate ≥98%
Supplier: Matrix Scientific
Synonyms:
1,1,1,3,3,3-Hexafluoroacetone sesquihydrate
004820-25G
004820-100G
004820-500G
101777-766EA
104.62
USD
101777-766
101777-764
101777-768
Hexafluoroacetone sesquihydrate ≥98%
Hexafluoroacetone sesquihydrate
Hazard: Toxic/Irritant
Formula:
C₃F₆O·1.5H₂O MW: 193.05 g/mol Melting Pt: 18…21 °C Density: 1.69 |
MDL Number:
MFCD06800792 UN: 2552 ADR: 6.1,II |
Specification Test Results
Melting point | 18-21° |
Density | 1.69 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...