1,8-Diiodo-3,3,4,4,5,5,6,6-octafluorooctane ≥97%

Supplier: Matrix Scientific

004787-1G
101777-754EA 127.69 USD
101777-754
1,8-Diiodo-3,3,4,4,5,5,6,6-octafluorooctane ≥97%
1,8-Diiodo-3,3,4,4,5,5,6,6-octafluorooctane

Hazard: Toxic

Formula: C₈H₈F₈I₂
Melting Pt: 91…93 °C
MDL Number: MFCD00153129
CAS Number: 2681-00-7

Order Now

Specification Test Results

Melting point 91-93°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR