Perfluoro-2-butene ≥97%
Supplier: Matrix Scientific
Synonyms:
FC-1318my
004781-100G
004781-25G
101777-748EA
1538.46
USD
101777-748
101777-746
Perfluoro-2-butene ≥97%
Perfluoro-2-butene
TCA listed: T
Hazard: Nonflammable Gas
Formula:
C₄F₈ MW: 200.03 g/mol Boiling Pt: 1.2 °C Melting Pt: –136…–134 °C |
CAS Number:
360-89-4 |
Specification Test Results
Boiling point | 1.2° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...