4-Bromobenzyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
α-Chloro-4-bromotoluene
, 1-(Chloromethyl)-4-bromobenzene
, 4-Bromo-α-chlorotoluene
, 1-Bromo-4-(chloromethyl)benzene
004763-25G
004763-5G
101777-744EA
100
USD
101777-744
102550-852
4-Bromobenzyl chloride ≥97%
4-Bromobenzyl chloride
Hazard: Corrosive/Lachrymator
Formula:
C₇H₆BrCl MW: 205.48 g/mol Boiling Pt: 106…107 °C (10 mmHg) Melting Pt: 39…40 °C |
MDL Number:
MFCD00040867 CAS Number: 589-17-3 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 106-107°/1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...