α,4-Dibromotoluene ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Bromobenzyl bromide
, 1-Bromo-4-(bromomethyl)benzene
004758-100G
101777-740EA
69.23
USD
101777-740
α,4-Dibromotoluene ≥98%
α,4-Dibromotoluene
TCA listed: T
Hazard: Corrosive/Lachrymator
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Melting Pt: 62…64 °C |
MDL Number:
MFCD00000179 CAS Number: 589-15-1 UN: 2811 ADR: 6.1,II |
Specification Test Results
Melting point | 62-64° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...