MATR004747

004747-500G 004747-100G
101777-730EA 70.62 USD
101777-730 101777-728
MATR004747
Purity: 99%
CAS: 98-16-8
MDL Number: MFCD00007797
Molecular Formula: C7H6F3N
Molecular Weight: 161,13
Boiling Point: 187°C
TCA listed: T
Hazard: Toxic/Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR