MATR004739

004739-100G
101777-718EA 45.77 USD
101777-718
MATR004739
Purity: 97%
CAS: 623-00-7
MDL Number: MFCD00001811
Molecular Formula: C7H4BrN
Molecular Weight: 182,03
Boiling Point: 235-237°C
Hazard: Toxic/Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR