Methyl pentafluoropropionate ≥99%

Supplier: Matrix Scientific

Synonyms: Methyl 2,2,3,3,3-pentafluoropropanoate

004700-100G
101777-702EA 65.55 USD
101777-702
Methyl pentafluoropropionate ≥99%
Methyl pentafluoropropionate

TCA listed: T
Hazard: Flammable

Formula: C₄H₃F₅O₂
MW: 178.06 g/mol
Boiling Pt: 59…60 °C
Density: 1.393
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Boiling point 59-60°
Density 1.393
ND20 1.2885

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR