3-Fluoroanisole ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Fluoro-3-methoxybenzene
004647-100G
004647-25G
101777-678EA
23.08
USD
101777-678
101777-680
3-Fluoroanisole ≥98%
3-Fluoroanisole
Hazard: Flammable
Formula:
C₇H₇FO MW: 126.13 g/mol Boiling Pt: 158 °C (743 mmHg) Density: 1.104 |
MDL Number:
MFCD00000335 CAS Number: 456-49-5 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 158°/743mm |
Density | 1.104 |
ND20 | 1.488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...