4-Fluoroanisole ≥99%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-4-methoxybenzene

004646-100G 004646-25G
101777-674EA 21.54 USD
101777-674 101777-676
4-Fluoroanisole ≥99%
4-Fluoroanisole

TCA listed: T
Hazard: Flammable

Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 157 °C
Melting Pt: –45 °C
Density: 1.114
MDL Number: MFCD00000348
CAS Number: 459-60-9
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Boiling point 157°
Density 1.114
ND20 1.488

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR