Ethyl bromodifluoroacetate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromo-2,2-difluoroacetateBromodifluoroacetic ethyl ester

004573-100G
101777-634EA 53.85 USD
101777-634
Ethyl bromodifluoroacetate ≥98%
Ethyl bromodifluoroacetate

Hazard: Flammable

Formula: BrF₂CCOOCH₂CH₃
MW: 202.98 g/mol
Boiling Pt: 112 °C (700 mmHg)
Density: 1.583
MDL Number: MFCD00042069
CAS Number: 667-27-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Boiling point 112°/700mm
Density 1.583
ND20 1.387

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR