Chlorotrifluoromethane ≥97%

Supplier: Matrix Scientific

004470-100G 004470-25G
101777-588EA 746.15 USD
101777-588 101777-590
Chlorotrifluoromethane ≥97%
Chlorotrifluoromethane

TCA listed: T
Hazard: Ozone Depleter/Nonflammable Gas

Formula: CClF₃
Boiling Pt: –81 °C
Melting Pt: –181 °C
CAS Number: 75-72-9

Order Now

Specification Test Results

Boiling point -81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR