Ethyl-2-fluoropropionate ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Fluoropropionic Acid Ethyl Ester
, Ethyl 2-fluoropropionate
004431-25G
004431-100G
004431-5G
102550-842EA
235.38
USD
102550-842
101777-558
102550-844
Ethyl-2-fluoropropionate ≥97%
Ethyl-2-fluoropropionate
Purity: 97%
CAS: 349-43-9
MDL Number: MFCD01320801
Molecular Formula: C5H9FO2
Molecular Weight: 120,12
Boiling Point: 122.5-123°C
Hazard: Flammable
Formula:
C₅H₉FO₂ MW: 120.12 g/mol Boiling Pt: 122.5…123 °C |
MDL Number:
MFCD01320801 CAS Number: 349-43-9 |
Specification Test Results
Boiling point | 122.5-123° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...