Perfluorodecanoic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanoic acid
004398-5G
101777-542EA
89.23
USD
101777-542
Perfluorodecanoic acid ≥98%
Perfluorodecanoic acid
TCA listed: T
Hazard: Toxic
Formula:
C₁₀HF₁₉O₂ MW: 514.09 g/mol Boiling Pt: 218 °C (740 mmHg) Melting Pt: 83…85 °C |
MDL Number:
MFCD00004175 CAS Number: 335-76-2 |
Specification Test Results
Boiling point | 218°/740mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...