Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yttrium(III) fluoride ≥99%

Supplier: Matrix Scientific

004365-100G
101777-524
Yttrium(III) fluoride ≥99%
Purity: 99+%
CAS: 13709-49-4
MDL Number: MFCD00049714
Molecular Formula: F3Y
Molecular Weight: 145,9
Hazard: Toxic


Formula: F₃Y
MW: 145,9 g/mol
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1387 °C
Density: 4.01 g/cm³ (20 °C)
MDL Number: MFCD00049714
CAS Number: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Melting point 1387°


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty