Xenon difluoride ≥99%

Supplier: Matrix Scientific

Synonyms: Xenon(II) fluoride

004362-5G
101777-520EA 497.95 USD
101777-520
Xenon difluoride ≥99%
Xenon difluoride

TCA listed: T
Hazard: Oxidizer/Irritant-Harmful/Stored under Argon

Formula: F₂Xe
MW: 169.29 g/mol
Melting Pt: 128 °C
Density: 4.32
MDL Number: MFCD00040538
CAS Number: 13709-36-9
UN: 3087
ADR: 5.1,II

Order Now

Specification Test Results

Melting point 128°
Density 4.32

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR