2-(Tributylstannyl)pyridine ≥90%

Supplier: Matrix Scientific

Synonyms: 2-(Tri-n-butylstannyl)pyridine

004351-5G
101777-516EA 89.23 USD
101777-516
2-(Tributylstannyl)pyridine ≥90%
2-(Tributylstannyl)pyridine

Balance is tributyl halide


Hazard: Toxic/Irritant/Refrigerate

Formula: C₁₇H₃₁NSn
MW: 368.15
Boiling Pt: 130…132 °C (0.8 mmHg)
Density: 1.14
MDL Number: MFCD00052052
CAS Number: 17997-47-6
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 130-132°/0
Density 1.14

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR