1-Naphthalenemethyl isothiocyanate ≥98%
Supplier: Matrix Scientific
004303-1G
004303-5G
101777-484EA
569.23
USD
101777-484
101777-482
1-Naphthalenemethyl isothiocyanate ≥98%
1-Naphthalenemethyl isothiocyanate
Hazard: Toxic/Refrigerate/Lachrymator
Formula:
C₁₂H₉NS Boiling Pt: 165 °C (2 mmHg) Melting Pt: 47…49 °C Density: 1.18 |
MDL Number:
MFCD00060346 CAS Number: 17112-82-2 |
Specification Test Results
Boiling point | 165°/2mm |
Density | 1.18 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...