2-(Dimethylamino)-6-fluorobenzonitrile ≥97%
Supplier: Matrix Scientific
004277-5G
004277-1G
101777-472EA
153.85
USD
101777-472
101777-470
2-(Dimethylamino)-6-fluorobenzonitrile ≥97%
2-(Dimethylamino)-6-fluorobenzonitrile
Hazard: Toxic
Formula:
C₉H₉FN₂ Boiling Pt: 131…135 °C |
MDL Number:
MFCD00052221 CAS Number: 96994-73-9 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 131-135° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...