Chlorocyclobutane ≥97%

Supplier: Matrix Scientific

Synonyms: Cyclobutyl chloride

004257-5G 004257-1G
101777-458EA 106.15 USD
101777-458 101777-460
Chlorocyclobutane ≥97%
Chlorocyclobutane

Hazard: Flammable

Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 83 °C
Density: 0.991
MDL Number: MFCD00013736
CAS Number: 1120-57-6
UN: 1127
ADR: 3,II

Order Now

Specification Test Results

Boiling point 83°
Density 0.991
ND20 1.436

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR