3-Bromoaniline ≥98%

Supplier: Matrix Scientific

004164-500G
101777-414EA 269.23 USD
101777-414
3-Bromoaniline ≥98%
3-Bromoaniline

TCA listed: T
Hazard: Toxic/Irritant-Harmful

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 251 °C
Melting Pt: 16.8 °C
Density: 1.59
Flash Pt: >110 °C
MDL Number: MFCD00007757
CAS Number: 591-19-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 251°
Density 1.58
ND20 1.625

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR