2,6-Xylyl isothiocyanate ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethylphenylisothiocyanate2,6-Dimethylphenyl isothiocyanate

004088-25G
101777-352EA 107.69 USD
101777-352
2,6-Xylyl isothiocyanate ≥98%
2,6-Xylyl isothiocyanate

Hazard: Toxic/Corrosive

Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 160 °C (35 mmHg)
Density: 1.08
MDL Number: MFCD00038715
CAS Number: 19241-16-8
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 160°/35mm
Density 1.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR