2,4-Dimethylphenethylamine ≥98%
Supplier: Matrix Scientific
004080-1G
004080-5G
101777-326EA
58.46
USD
101777-326
101777-328
2,4-Dimethylphenethylamine ≥98%
2,4-Dimethylphenethylamine
Hazard: Corrosive/Irritant-Harmful
Specification Test Results
Boiling point | 95°/10mm |
Density | 0.95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...